siRNA Details
| Gene Symbol: HYAL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012269.2 | NP_036401.2 | 3 | 1174 | ||
| XM_011515990.2 | XP_011514292.1 | 3 | 1250 | |||
| XM_011515991.2 | XP_011514293.1 | 3 | 832 | |||
| XM_017011911.1 | XP_016867400.1 | 4 | 877 | |||
| GenBank mRNA | AB470346.1 | - | 2 | 606 | ||
| AF009010.1 | - | 3 | 1177 | |||
| BC104788.1 | - | 2 | 612 | 8143791 | ||
| BC104790.1 | - | 2 | 612 | 8143793 | ||

HYAL4
Genomic Map