siRNA Details
Gene Symbol: CHST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024533.4 | NP_078809.2 | 3 | 2902 | ||
GenBank mRNA | AF246718.1 | - | 3 | 2903 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CHST5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024533.4 | NP_078809.2 | 3 | 2902 | ||
GenBank mRNA | AF246718.1 | - | 3 | 2903 |