siRNA Details
| Gene Symbol: DDAH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001303007.1 | NP_001289936.1 | 5 | 1714 | ||
| NM_001303008.1 | NP_001289937.1 | 6 | 814 | |||
| NM_013974.2 | NP_039268.1 | 6 | 889 | |||
| XM_011514448.2 | XP_011512750.1 | 6 | 965 | |||
| GenBank mRNA | AF070667.1 | - | 6 | 886 | ||
| AF087894.1 | - | 6 | 886 | |||
| AK026191.1 | - | 5 | 1241 | |||
| AK098671.1 | - | 6 | 876 | |||
| BC001435.2 | - | 6 | 801 | 3138832 | ||
| EB387244.1 | - | 6 | 788 | |||
| FJ224290.1 | - | 6 | 886 | |||

DDAH2
Genomic Map