siRNA Details
| Gene Symbol: PADI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012387.2 | NP_036519.2 | 14 | 1641 | ||
| XM_011541150.1 | XP_011539452.1 | 12 | 1453 | |||
| XM_011541151.1 | XP_011539453.1 | 12 | 1341 | |||
| XM_011541152.1 | XP_011539454.1 | 11 | 1173 | |||
| XM_011541157.1 | XP_011539459.1 | 11 | 1120 | |||
| GenBank mRNA | AB017919.1 | - | 14 | 1639 | ||
| AK290507.1 | - | 14 | 1639 | |||
| AK314839.1 | - | 14 | 1641 | |||
| BC025718.1 | - | 14 | 1625 | 5228777 | ||

PADI4
Genomic Map