siRNA Details
| Gene Symbol: PDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321978.1 | NP_001308907.1 | 6 | 848 | ||
| NM_001321979.1 | NP_001308908.1 | 6 | 931 | |||
| NM_014317.4 | NP_055132.2 | 6 | 848 | |||
| XM_011519437.2 | XP_011517739.1 | 3 | 1248 | |||
| XM_017016011.1 | XP_016871500.1 | 4 | 326 | |||
| GenBank mRNA | AB209763.1 | - | 5 | 2886 | ||
| AB210838.1 | - | 6 | 606 | |||
| AF118395.1 | - | 5 | 468 | |||
| BC049211.1 | - | 6 | 558 | |||
| BC063635.1 | - | 6 | 576 | 4397753 | ||

PDSS1
Genomic Map