siRNA Details
| Gene Symbol: LEMD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167614.1 | NP_001161086.1 | 9 | 2238 | ||
| NM_014319.4 | NP_055134.2 | 9 | 2241 | |||
| GenBank mRNA | AF112299.2 | - | 9 | 2221 | ||
| AK024954.1 | - | 9 | 916 | |||
| AK295406.1 | - | 3 | 289 | |||
| AK312473.1 | - | 9 | 2238 | |||
| AL137533.1 | - | 1 | 77 | |||

LEMD3
Genomic Map