siRNA Details
| Gene Symbol: CLEC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301167.1 | NP_001288096.1 | 4 | 447 | ||
| NM_013252.2 | NP_037384.1 | 5 | 492 | |||
| XM_011515995.2 | XP_011514297.1 | 3 | 379 | |||
| XM_017011915.1 | XP_016867404.1 | 4 | 450 | |||
| XM_017011916.1 | XP_016867405.1 | 5 | 518 | |||
| XM_017011917.1 | XP_016867406.1 | 4 | 449 | |||
| GenBank mRNA | AF139768.1 | - | 5 | 446 | ||
| AJ271684.1 | - | 5 | 492 | |||
| AK292766.1 | - | 5 | 481 | |||
| AK295200.1 | - | 4 | 384 | |||
| AK313703.1 | - | 5 | 481 | |||
| BC093714.1 | - | 4 | 316 | 7939559 | ||
| BC112099.1 | - | 4 | 316 | 8327567 | ||
| BC113098.1 | - | 5 | 434 | 40076723 | ||
| BC113099.1 | - | 4 | 365 | 40076725 | ||
| BC143392.1 | - | 3 | 247 | |||

CLEC5A
Genomic Map