siRNA Details
| Gene Symbol: CLEC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301167.1 | NP_001288096.1 | 2 | 277 | ||
| NM_013252.2 | NP_037384.1 | 2 | 253 | |||
| XM_011515995.2 | XP_011514297.1 | 2 | 269 | |||
| XM_017011915.1 | XP_016867404.1 | 2 | 271 | |||
| XM_017011916.1 | XP_016867405.1 | 2 | 279 | |||
| XM_017011917.1 | XP_016867406.1 | 2 | 279 | |||
| GenBank mRNA | AF139768.1 | - | 2 | 207 | ||
| AJ271684.1 | - | 2 | 253 | |||
| AK292766.1 | - | 2 | 242 | |||
| AK295200.1 | - | 2 | 214 | |||
| AK307095.1 | - | 2 | 277 | |||
| AK313703.1 | - | 2 | 242 | |||
| BC093714.1 | - | 1 | 77 | 7939559 | ||
| BC112099.1 | - | 1 | 77 | 8327567 | ||
| BC113098.1 | - | 2 | 195 | 40076723 | ||
| BC113099.1 | - | 2 | 195 | 40076725 | ||
| BC143392.1 | - | 1 | 77 | |||

CLEC5A
Genomic Map