siRNA Details
| Gene Symbol: CLEC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301167.1 | NP_001288096.1 | 5 | 556 | ||
| NM_013252.2 | NP_037384.1 | 6 | 601 | |||
| XM_011515995.2 | XP_011514297.1 | 4 | 488 | |||
| XM_017011915.1 | XP_016867404.1 | 5 | 559 | |||
| XM_017011916.1 | XP_016867405.1 | 6 | 627 | |||
| XM_017011917.1 | XP_016867406.1 | 5 | 558 | |||
| GenBank mRNA | AF139768.1 | - | 6 | 552 | ||
| AJ271684.1 | - | 6 | 601 | |||
| AK292766.1 | - | 6 | 590 | |||
| AK295200.1 | - | 5 | 493 | |||
| AK307095.1 | - | 4 | 419 | |||
| AK313703.1 | - | 6 | 590 | |||
| BC093714.1 | - | 5 | 425 | 7939559 | ||
| BC112099.1 | - | 5 | 425 | 8327567 | ||
| BC113098.1 | - | 6 | 543 | 40076723 | ||
| BC113099.1 | - | 5 | 474 | 40076725 | ||
| BC143392.1 | - | 4 | 356 | |||

CLEC5A
Genomic Map