siRNA Details
| Gene Symbol: SH3BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018957.3 | NP_061830.3 | 9 | 824 | ||
| GenBank mRNA | AK124370.1 | - | 10 | 1651 | ||
| AK126556.1 | - | 8 | 1519 | |||
| AK301163.1 | - | 9 | 806 | |||
| AY364241.1 | - | 6 | 521 | |||
| BC008282.1 | - | 7 | 617 | 3639782 | ||
| CR456576.1 | - | 9 | 743 | |||

SH3BP1
Genomic Map