siRNA Details
| Gene Symbol: PLXNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012401.3 | NP_036533.2 | 3 | 1304 | ||
| XM_005261909.1 | XP_005261966.1 | 4 | 1352 | |||
| XM_005261910.2 | XP_005261967.1 | 4 | 1363 | |||
| XM_005261911.4 | XP_005261968.1 | 3 | 1302 | |||
| XM_006724413.2 | XP_006724476.1 | 5 | 1422 | |||
| XM_011530682.2 | XP_011528984.1 | 4 | 1503 | |||
| XM_017028703.1 | XP_016884192.1 | 5 | 1403 | |||
| XM_017028704.1 | XP_016884193.1 | 4 | 1356 | |||
| GenBank mRNA | AB002313.1 | - | 2 | 1221 | ||
| BC127770.1 | - | 4 | 1408 | |||

PLXNB2
Genomic Map