siRNA Details
| Gene Symbol: PLXNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012401.3 | NP_036533.2 | 8 | 1906 | ||
| XM_005261909.1 | XP_005261966.1 | 9 | 1954 | |||
| XM_005261910.2 | XP_005261967.1 | 9 | 1965 | |||
| XM_005261911.4 | XP_005261968.1 | 8 | 1904 | |||
| XM_006724413.2 | XP_006724476.1 | 10 | 2024 | |||
| XM_011530682.2 | XP_011528984.1 | 9 | 2105 | |||
| XM_017028703.1 | XP_016884192.1 | 10 | 2005 | |||
| XM_017028704.1 | XP_016884193.1 | 9 | 1958 | |||
| GenBank mRNA | AB002313.1 | - | 7 | 1823 | ||

PLXNB2
Genomic Map