siRNA Details
| Gene Symbol: LSM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130710.1 | NP_001124182.1 | 2 | 78 | ||
| NM_001139499.1 | NP_001132971.1 | 2 | 317 | |||
| NM_012322.2 | NP_036454.1 | 2 | 141 | |||
| NR_024466.1 | 2 | 888 | ||||
| GenBank mRNA | AF182291.1 | - | 2 | 102 | ||
| AJ238097.1 | - | 2 | 95 | |||
| AK024217.1 | - | 2 | 124 | |||
| BC005938.1 | - | 2 | 101 | 4132686 | ||
| BF667238.1 | - | 2 | 142 | |||
| BF700164.1 | - | 2 | 78 | |||
| DA736378.1 | - | 2 | 308 | |||

LSM5
Genomic Map