siRNA Details
| Gene Symbol: SMPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014332.2 | NP_055147.1 | 2, 3 | 285 | ||
| NR_045617.1 | 2, 3 | 285 | ||||
| GenBank mRNA | AF129505.1 | - | 2, 3 | 237 | ||
| AJ250584.1 | - | 2, 3 | 237 | |||
| AK312134.1 | - | 2, 3 | 285 | |||
| BC005948.1 | - | 2, 3 | 166 | 4246501 | ||
| BF126275.1 | - | 2, 3 | 147 | |||

SMPX
Genomic Map