siRNA Details
| Gene Symbol: SHPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013276.2 | NP_037408.2 | 2 | 285 | ||
| GenBank mRNA | AF163573.1 | - | 2 | 285 | ||
| AK055609.1 | - | 2 | 211 | |||
| AK298360.1 | - | 2 | 233 | |||
| AK312428.1 | - | 2 | 232 | |||
| AL832420.1 | - | 2 | 224 | |||
| BC020543.1 | - | 2 | 241 | 4413491 | ||

SHPK
Genomic Map