siRNA Details
| Gene Symbol: FLRT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013280.4 | NP_037412.2 | 2 | 1568 | ||
| XM_005273861.1 | XP_005273918.1 | 3 | 2454 | |||
| XM_011544883.1 | XP_011543185.1 | 2 | 1408 | |||
| GenBank mRNA | AF169675.1 | - | 1 | 1227 | ||
| AK289931.1 | - | 2 | 1885 | |||
| AY358308.1 | - | 2 | 1689 | |||
| BC018370.1 | - | 2 | 1568 | 4214494 | ||

FLRT1
Genomic Map