siRNA Details
| Gene Symbol: ZNF318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014345.2 | NP_055160.2 | 6 | 3028 | ||
| XM_011514451.2 | XP_011512753.1 | 6 | 2629 | |||
| XM_011514452.2 | XP_011512754.1 | 6 | 3261 | |||
| GenBank mRNA | AK025505.1 | - | 3 | 1533 | ||
| BC030687.1 | - | 6 | 3063 | |||
| BC098434.1 | - | 6 | 3003 | |||

ZNF318
Genomic Map