siRNA Details
| Gene Symbol: SHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012435.2 | NP_036567.2 | 12 | 1633 | ||
| XM_011527895.2 | XP_011526197.1 | 11 | 1513 | |||
| GenBank mRNA | AB001451.1 | - | 12 | 1507 | ||
| AL360254.1 | - | 10 | 1063 | |||
| BC048290.1 | - | 2 | 335 | |||
| BY796046.2 | - | 3 | 450 | |||
| CF457169.1 | - | 3 | 435 | |||

SHC2
Genomic Map