siRNA Details
| Gene Symbol: CBY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002880.1 | NP_001002880.2 | 6 | 610 | ||
| NM_015373.3 | NP_056188.1 | 5 | 470 | |||
| GenBank mRNA | AB015347.1 | - | 2 | 157 | ||
| AB111855.1 | - | 4 | 348 | |||
| AF331041.1 | - | 5 | 406 | |||
| AK311928.1 | - | 5 | 483 | |||
| AL050345.1 | - | 5 | 449 | |||
| AL136686.1 | - | 5 | 453 | |||
| BC016139.1 | - | 5 | 448 | 3922707 | ||
| CR456410.1 | - | 4 | 362 | |||

CBY1
Genomic Map