siRNA Details
| Gene Symbol: CFAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012337.2 | NP_036469.2 | 6 | 699 | ||
| GenBank mRNA | AF094758.1 | - | 6 | 732 | ||
| AK225566.1 | - | 6 | 745 | |||
| AK225582.1 | - | 6 | 745 | |||
| AK292249.1 | - | 6 | 748 | |||
| AK308262.1 | - | 6 | 742 | |||
| BC027874.1 | - | 6 | 699 | |||
| BC089391.1 | - | 6 | 894 | 30387473 | ||
| BC109055.1 | - | 5 | 674 | |||

CFAP45
Genomic Map