siRNA Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012413.3 | NP_036545.1 | 7 | 1158 | ||
| GenBank mRNA | AK290605.1 | - | 7 | 1140 | ||
| BC020456.1 | - | 7 | 1059 | |||
| BC036721.1 | - | 7 | 1037 | 5224222 | ||
| BC047756.2 | - | 7 | 1135 | 6058335 | ||
| X71125.1 | - | 7 | 1047 | |||

QPCT
Genomic Map