siRNA Details
| Gene Symbol: BRI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015379.4 | NP_056194.1 | 3 | 698 | ||
| GenBank mRNA | AB055977.1 | - | 3 | 690 | ||
| AF041430.2 | - | 3 | 668 | |||
| AF106966.1 | - | 3 | 649 | |||
| BC018737.1 | - | 3 | 690 | 4875257 | ||
| BC062370.1 | - | 3 | 662 | 6598528 | ||

BRI3
Genomic Map