siRNA Details
| Gene Symbol: HECTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015382.3 | NP_056197.3 | 6 | 1632 | ||
| XM_005267502.2 | XP_005267559.2 | 6 | 1794 | |||
| XM_011536619.2 | XP_011534921.1 | 6 | 1230 | |||
| XM_011536621.2 | XP_011534923.1 | 6 | 1230 | |||
| XM_017021148.1 | XP_016876637.1 | 5 | 1124 | |||
| XM_017021149.1 | XP_016876638.1 | 5 | 1124 | |||
| XM_017021150.1 | XP_016876639.1 | 5 | 1124 | |||
| XM_017021151.1 | XP_016876640.1 | 6 | 1826 | |||
| XM_017021152.1 | XP_016876641.1 | 6 | 1826 | |||
| XM_017021153.1 | XP_016876642.1 | 6 | 1230 | |||
| GenBank mRNA | AY254380.1 | - | 6 | 1426 | ||

HECTD1
Genomic Map