siRNA Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308053.1 | NP_001294982.1 | 5 | 576 | ||
| NM_014353.4 | NP_055168.2 | 4 | 537 | |||
| XM_011522448.1 | XP_011520750.1 | 4 | 563 | |||
| XM_011522450.2 | XP_011520752.1 | 3 | 1354 | |||
| GenBank mRNA | AB027137.1 | - | 3 | 208 | ||
| AF498952.1 | - | 3 | 205 | |||
| AK308560.1 | - | 6 | 633 | |||
| AK309666.1 | - | 5 | 541 | |||
| AK314110.1 | - | 5 | 576 | |||
| AY646153.1 | - | 4 | 554 | |||
| BC066913.1 | - | 4 | 543 | 5262795 | ||

RAB26
Genomic Map