siRNA Details
| Gene Symbol: ABTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145804.2 | NP_665803.2 | 3 | 1656 | ||
| GenBank mRNA | AK095632.1 | - | 3 | 903 | ||
| AK291744.1 | - | 3 | 1657 | |||
| BC093968.1 | - | 3 | 768 | 7939813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABTB2| Gene Symbol: ABTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145804.2 | NP_665803.2 | 3 | 1656 | ||
| GenBank mRNA | AK095632.1 | - | 3 | 903 | ||
| AK291744.1 | - | 3 | 1657 | |||
| BC093968.1 | - | 3 | 768 | 7939813 | ||