siRNA Details
| Gene Symbol: TECPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015395.2 | NP_056210.1 | 8, 9 | 1236 | ||
| XM_005250253.3 | XP_005250310.1 | 8, 9 | 1208 | |||
| XM_017011938.1 | XP_016867427.1 | 8, 9 | 1220 | |||
| GenBank mRNA | AB037779.1 | - | 6, 7 | 812 | ||
| AK057048.1 | - | 7, 8 | 963 | |||
| AK292996.1 | - | 8, 9 | 1138 | |||
| BC053591.1 | - | 8, 9 | 1141 | 5502429 | ||

TECPR1
Genomic Map