siRNA Details
| Gene Symbol: WHRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083885.2 | NP_001077354.2 | 11 | 2018 | ||
| NM_001173425.1 | NP_001166896.1 | 11 | 3179 | |||
| NM_015404.3 | NP_056219.3 | 11 | 3182 | |||
| XM_011518484.1 | XP_011516786.1 | 12 | 3268 | |||
| XM_011518485.1 | XP_011516787.1 | 13 | 3116 | |||
| XM_011518486.1 | XP_011516788.1 | 12 | 3265 | |||
| XM_011518487.1 | XP_011516789.1 | 11 | 3142 | |||
| XM_011518488.1 | XP_011516790.1 | 10 | 3025 | |||
| XM_011518495.1 | XP_011516797.1 | 9 | 1520 | |||
| GenBank mRNA | AB040959.1 | - | 11 | 2696 | ||
| AK022854.1 | - | 7 | 1896 | |||
| AL110228.1 | - | 11 | 1976 | |||
| BC011918.2 | - | 2 | 291 | |||
| BC014524.1 | - | 3679 | ||||
| BC136416.1 | - | 11 | 2928 | 9020404 | ||
| BC142614.1 | - | 11 | 3182 | 40147287 | ||
| BC142684.1 | - | 11 | 3157 | 40148465 | ||

WHRN
Genomic Map