siRNA Details
| Gene Symbol: ABHD14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015407.4 | NP_056222.2 | 3 | 423 | ||
| GenBank mRNA | AK308592.1 | - | 3 | 347 | ||
| AL050015.1 | - | 3 | 334 | |||
| AY358201.1 | - | 3 | 380 | |||
| BC002571.2 | - | 3 | 381 | 3143135 | ||
| CN256406.1 | - | 3 | 427 | |||
| Gene Symbol: ABHD14A-ACY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316331.1 | NP_001303260.1 | 3 | 423 | ||
| GenBank mRNA | AK298086.1 | - | 3 | 399 | ||

ABHD14A
Genomic Map