siRNA Details
| Gene Symbol: NEPRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319109.1 | NP_001306038.1 | 7 | 1371 | ||
| NM_001319110.1 | NP_001306039.1 | 6 | 1216 | |||
| NM_001319111.1 | NP_001306040.1 | 8 | 1185 | |||
| NM_001319112.1 | NP_001306041.1 | 8 | 1384 | |||
| NM_001319114.1 | NP_001306043.1 | 8 | 1430 | |||
| NM_001319115.1 | NP_001306044.1 | 8 | 1426 | |||
| NM_015412.3 | NP_056227.2 | 9 | 1556 | |||
| NR_134969.1 | 6 | 1309 | ||||
| XM_005247276.2 | XP_005247333.1 | 9 | 1523 | |||
| XM_011512635.2 | XP_011510937.1 | 9 | 1278 | |||
| XM_011512636.2 | XP_011510938.1 | 9 | 1282 | |||
| XM_011512638.2 | XP_011510940.1 | 8 | 1177 | |||
| XM_017006102.1 | XP_016861591.1 | 9 | 1545 | |||
| XM_017006103.1 | XP_016861592.1 | 8 | 1440 | |||
| GenBank mRNA | AK024325.1 | - | 9 | 1514 | ||
| AK295165.1 | - | 8 | 1315 | |||
| AK295954.1 | - | 6 | 1153 | |||
| AK300811.1 | - | 8 | 1118 | |||
| AK301161.1 | - | 8 | 1344 | |||
| AK304022.1 | - | 3 | 644 | |||
| AL117573.1 | - | 9 | 1498 | |||
| BC014673.2 | - | 8 | 2014 | |||
| BC067743.1 | - | 9 | 1516 | 4812681 | ||
| CR749341.1 | - | 7 | 1372 | |||

NEPRO
Genomic Map