siRNA Details
| Gene Symbol: NEPRO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001319109.1 | NP_001306038.1 | 4 | 612 | ||
| NM_001319110.1 | NP_001306039.1 | 3 | 457 | |||
| NM_001319112.1 | NP_001306041.1 | 5 | 625 | |||
| NM_001319114.1 | NP_001306043.1 | 5 | 671 | |||
| NM_001319115.1 | NP_001306044.1 | 5 | 667 | |||
| NM_015412.3 | NP_056227.2 | 6 | 797 | |||
| NR_134969.1 | 4 | 612 | ||||
| XM_005247276.2 | XP_005247333.1 | 6 | 764 | |||
| XM_017006102.1 | XP_016861591.1 | 6 | 760 | |||
| XM_017006103.1 | XP_016861592.1 | 5 | 655 | |||
| GenBank mRNA | AK024325.1 | - | 6 | 755 | ||
| AK293218.1 | - | 4 | 550 | |||
| AK295165.1 | - | 5 | 556 | |||
| AK295954.1 | - | 3 | 394 | |||
| AK301161.1 | - | 5 | 585 | |||
| AK304083.1 | - | 5 | 600 | |||
| AL117573.1 | - | 6 | 739 | |||
| AL537045.3 | - | 5 | 592 | |||
| BC014673.2 | - | 5 | 1255 | |||
| BC067743.1 | - | 6 | 757 | 4812681 | ||
| CR749341.1 | - | 4 | 613 | |||

NEPRO
Genomic Map