siRNA Details
| Gene Symbol: TMEM186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015421.3 | NP_056236.2 | 2 | 623 | ||
| GenBank mRNA | AK093383.1 | - | 1 | 1758 | ||
| AK225638.1 | - | 2 | 605 | |||
| AK314404.1 | - | 2 | 614 | |||
| AL080088.1 | - | 2 | 612 | |||
| BC015912.1 | - | 1 | 598 | 4554816 | ||

TMEM186
Genomic Map