siRNA Details
| Gene Symbol: POC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161580.1 | NP_001155052.1 | 9 | 1232 | ||
| NM_001161581.1 | NP_001155053.1 | 9 | 1303 | |||
| NM_015426.4 | NP_056241.3 | 9 | 1232 | |||
| XM_017006104.1 | XP_016861593.1 | 9 | 1025 | |||
| GenBank mRNA | AK075289.1 | - | 9 | 947 | ||
| AK315688.1 | - | 9 | 954 | |||
| AL117629.1 | - | 4 | 271 | |||
| BC007417.2 | - | 8 | 882 | |||
| BC110877.1 | - | 9 | 1232 | 5588109 | ||
| BC119692.2 | - | 9 | 942 | 40116721 | ||
| BC119693.2 | - | 9 | 1018 | |||

POC1A
Genomic Map