siRNA Details
| Gene Symbol: ZNF473 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 2943 | ||
| GenBank mRNA | AB032967.1 | - | 5 | 1700 | ||
| AF454744.1 | - | 4 | 1655 | |||
| AK054671.1 | - | 1 | 842 | |||
| AK292452.1 | - | 5 | 1593 | |||
| AK302293.1 | - | 4 | 1550 | |||
| BC018612.1 | - | 5 | 1623 | 4634056 | ||
| BX648093.1 | - | 5 | 1744 | |||

ZNF473
Genomic Map