siRNA Details
| Gene Symbol: TRIM58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015431.3 | NP_056246.3 | 6 | 1389 | ||
| GenBank mRNA | AF327057.1 | - | 6 | 1389 | ||
| AK096188.1 | - | 6 | 1271 | |||
| BC074748.2 | - | 5 | 933 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM58| Gene Symbol: TRIM58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015431.3 | NP_056246.3 | 6 | 1389 | ||
| GenBank mRNA | AF327057.1 | - | 6 | 1389 | ||
| AK096188.1 | - | 6 | 1271 | |||
| BC074748.2 | - | 5 | 933 | |||