siRNA Details
| Gene Symbol: PLEKHG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129727.2 | NP_001123199.1 | 15 | 2592 | ||
| NM_001129728.1 | NP_001123200.1 | 14 | 2552 | |||
| NM_001129729.2 | NP_001123201.1 | 14 | 2525 | |||
| NM_001129731.2 | NP_001123203.1 | 12 | 2531 | |||
| XM_011522985.2 | XP_011521287.1 | 14 | 2522 | |||
| XM_011522986.2 | XP_011521288.1 | 14 | 2654 | |||
| XM_011522987.2 | XP_011521289.1 | 15 | 2598 | |||
| XM_011522988.2 | XP_011521290.1 | 14 | 2471 | |||
| GenBank mRNA | AB197663.1 | - | 13 | 4788 | ||
| AB197664.1 | - | 12 | 4955 | |||
| AK024475.1 | - | 14 | 2487 | |||
| AK304723.1 | - | 12 | 1943 | |||
| AK308298.1 | - | 14 | 2461 | |||
| AL117435.1 | - | 9 | 1424 | |||
| BC054486.1 | - | 12 | 2466 | 4820741 | ||
| BC063501.1 | - | 14 | 2463 | 5527705 | ||
| BC082974.1 | - | 15 | 2560 | 6291175 | ||

PLEKHG4
Genomic Map