siRNA Details
| Gene Symbol: CCDC28A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015439.2 | NP_056254.1 | 2 | 411 | ||
| XM_011535728.2 | XP_011534030.1 | 2 | 414 | |||
| GenBank mRNA | AJ420568.1 | - | 2 | 162 | ||
| AK300672.1 | - | 2 | 162 | |||
| AL050197.1 | - | 2 | 161 | |||
| AY167571.1 | - | 2 | 429 | |||
| BC004464.1 | - | 2 | 145 | 2820737 | ||
| BC013019.2 | - | 2 | 143 | |||
| BC080566.1 | - | 2 | 145 | |||
| BC108717.1 | - | 2 | 334 | 6068596 | ||

CCDC28A
Genomic Map