siRNA Details
| Gene Symbol: CCDC28A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015439.2 | NP_056254.1 | 2 | 469 | ||
| XM_011535728.2 | XP_011534030.1 | 2 | 472 | |||
| GenBank mRNA | AJ420568.1 | - | 2 | 220 | ||
| AK300672.1 | - | 2 | 220 | |||
| AL050197.1 | - | 2 | 219 | |||
| AY167571.1 | - | 2 | 487 | |||
| BC004464.1 | - | 2 | 203 | 2820737 | ||
| BC013019.2 | - | 2 | 201 | |||
| BC080566.1 | - | 2 | 203 | |||
| BC108717.1 | - | 2 | 392 | 6068596 | ||

CCDC28A
Genomic Map