siRNA Details
| Gene Symbol: NDUFAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199069.1 | NP_951032.1 | 5 | 1110 | ||
| NM_199070.1 | NP_951033.1 | 5 | 673 | |||
| NM_199073.1 | NP_951047.1 | 5 | 744 | |||
| NM_199074.1 | NP_951056.1 | 5 | 625 | |||
| GenBank mRNA | AL049955.1 | - | 5 | 677 | ||
| BC002873.2 | - | 5 | 660 | 3945015 | ||
| BC111004.1 | - | 5 | 1740 | 5744973 | ||
| BQ652614.1 | - | 5 | 625 | |||

NDUFAF3
Genomic Map