siRNA Details
| Gene Symbol: MYRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284423.1 | NP_001271352.1 | 11 | 1863 | ||
| NM_001284424.1 | NP_001271353.1 | 11 | 2052 | |||
| NM_001284425.1 | NP_001271354.1 | 10 | 1915 | |||
| NM_001284426.1 | NP_001271355.1 | 9 | 1673 | |||
| NM_015460.3 | NP_056275.2 | 11 | 2052 | |||
| NR_104316.1 | 11 | 2052 | ||||
| XM_011533575.1 | XP_011531877.1 | 11 | 2024 | |||
| GenBank mRNA | AB083783.1 | - | 10 | 1765 | ||
| AF396687.1 | - | 11 | 1901 | |||
| AK125334.1 | - | 9 | 1539 | |||
| AK126013.1 | - | 11 | 2006 | |||
| AK294714.1 | - | 9 | 1663 | |||
| AK312412.1 | - | 11 | 1862 | |||
| AK316068.1 | - | 9 | 1673 | |||
| BC092512.1 | - | 11 | 2051 | 3093499 | ||
| BC109311.1 | - | 10 | 1743 | 40036096 | ||
| BC109312.1 | - | 11 | 1880 | 40036097 | ||

MYRIP
Genomic Map