siRNA Details
| Gene Symbol: NIPSNAP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015469.1 | NP_056284.1 | 3 | 491 | ||
| GenBank mRNA | AF161417.1 | - | 3 | 472 | ||
| AK024015.1 | - | 3 | 452 | |||
| AK301410.1 | - | 3 | 482 | |||
| AL117557.1 | - | 3 | 435 | |||
| BC005935.1 | - | 3 | 491 | 4064889 | ||
| BC045830.1 | - | 3 | 431 | |||

NIPSNAP3A
Genomic Map