siRNA Details
| Gene Symbol: NIPSNAP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015469.1 | NP_056284.1 | 2 | 210 | ||
| GenBank mRNA | AF161417.1 | - | 2 | 191 | ||
| AK024015.1 | - | 2 | 171 | |||
| AK301410.1 | - | 2 | 201 | |||
| AL117557.1 | - | 2 | 154 | |||
| BC005935.1 | - | 2 | 210 | 4064889 | ||
| BC045830.1 | - | 2 | 150 | |||

NIPSNAP3A
Genomic Map