siRNA Details
| Gene Symbol: TMEM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110503.2 | NP_001103973.1 | 3 | 411 | ||
| NM_001286487.1 | NP_001273416.1 | 2 | 350 | |||
| NM_015497.4 | NP_056312.2 | 3 | 411 | |||
| XM_005254287.1 | XP_005254344.1 | 3 | 262 | |||
| XM_006720468.2 | XP_006720531.1 | 3 | 267 | |||
| XM_011521443.2 | XP_011519745.1 | 2 | 179 | |||
| XM_011521444.2 | XP_011519746.1 | 3 | 262 | |||
| XM_017022067.1 | XP_016877556.1 | 3 | 267 | |||
| GenBank mRNA | AF132733.1 | - | 3 | 267 | ||
| AK074870.1 | - | 3 | 260 | |||
| AK075403.1 | - | 3 | 297 | |||
| AK289591.1 | - | 3 | 260 | |||
| AK290150.1 | - | 3 | 246 | |||
| AK290484.1 | - | 2 | 194 | |||
| AK297816.1 | - | 3 | 256 | |||
| AK309915.1 | - | 3 | 260 | |||
| AK309954.1 | - | 2 | 200 | |||
| BC005335.1 | - | 3 | 263 | |||
| DC341748.1 | - | 3 | 384 | |||
| HY036655.1 | - | 3 | 411 | |||

TMEM87A
Genomic Map