siRNA Details
| Gene Symbol: SH2B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145795.1 | NP_001139267.1 | 4 | 1328 | ||
| NM_001145796.1 | NP_001139268.1 | 4 | 1328 | |||
| NM_001145797.1 | NP_001139269.1 | 4 | 1328 | |||
| NM_001145812.1 | NP_001139284.1 | 4 | 1427 | |||
| NM_001308293.1 | NP_001295222.1 | 6 | 1584 | |||
| NM_001308294.1 | NP_001295223.1 | 3 | 263 | |||
| NM_015503.2 | NP_056318.2 | 3 | 3158 | |||
| XM_017023114.1 | XP_016878603.1 | 3 | 1747 | |||
| XM_017023115.1 | XP_016878604.1 | 5 | 1454 | |||
| XM_017023116.1 | XP_016878605.1 | 4 | 2812 | |||
| XM_017023117.1 | XP_016878606.1 | 3 | 1747 | |||
| XM_017023118.1 | XP_016878607.1 | 3 | 434 | |||
| XM_017023119.1 | XP_016878608.1 | 3 | 263 | |||
| XM_017023120.1 | XP_016878609.1 | 2 | 74 | |||
| GenBank mRNA | AB037720.1 | - | 3 | 4347 | ||
| AF227967.1 | - | 4 | 1184 | |||
| AF227968.1 | - | 4 | 1184 | |||
| AF227969.1 | - | 4 | 1184 | |||
| AK027488.1 | - | 3 | 493 | |||
| AK055104.1 | - | 6 | 1495 | |||
| AK290332.1 | - | 4 | 1427 | |||
| AK297080.1 | - | 3 | 214 | |||
| AK298569.1 | - | 3 | 313 | |||
| AL049924.1 | - | 3 | 628 | |||
| AL713760.1 | - | 3 | 3158 | |||
| BC010704.1 | - | 3 | 881 | 3866324 | ||
| DB026545.1 | - | 3 | 263 | |||

SH2B1
Genomic Map