siRNA Details
| Gene Symbol: CHMP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244644.1 | NP_001231573.1 | 2 | 381 | ||
| NM_014043.3 | NP_054762.2 | 3 | 473 | |||
| XM_011533576.2 | XP_011531878.1 | 3 | 304 | |||
| GenBank mRNA | AF151842.1 | - | 3 | 432 | ||
| AK002165.1 | - | 3 | 446 | |||
| AK002180.1 | - | 3 | 451 | |||
| AK222654.1 | - | 3 | 433 | |||
| AK296072.1 | - | 2 | 373 | |||
| AK316585.1 | - | 3 | 437 | |||
| AL080122.1 | - | 3 | 446 | |||
| BC001553.1 | - | 3 | 289 | 3460712 | ||
| BI821848.1 | - | 3 | 450 | |||
| BM828410.1 | - | 3 | 473 | |||
| CR533456.1 | - | 3 | 213 | |||
| DA426159.1 | - | 3 | 444 | |||

CHMP2B
Genomic Map