siRNA Details
| Gene Symbol: HINFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243259.1 | NP_001230188.1 | 6, 7 | 881 | ||
| NM_015517.4 | NP_056332.2 | 7, 8 | 998 | |||
| NM_198971.2 | NP_945322.1 | 6, 7 | 881 | |||
| XM_011542744.2 | XP_011541046.1 | 6, 7 | 990 | |||
| XM_011542745.2 | XP_011541047.1 | 6, 7 | 840 | |||
| XM_011542746.1 | XP_011541048.1 | 5, 6 | 661 | |||
| XM_017017499.1 | XP_016872988.1 | 6, 7 | 938 | |||
| XM_017017500.1 | XP_016872989.1 | 6, 7 | 817 | |||
| XM_017017501.1 | XP_016872990.1 | 7, 8 | 944 | |||
| XM_017017502.1 | XP_016872991.1 | 9, 10 | 1269 | |||
| XM_017017503.1 | XP_016872992.1 | 7, 8 | 1034 | |||
| XM_017017504.1 | XP_016872993.1 | 7, 8 | 909 | |||
| XM_017017505.1 | XP_016872994.1 | 7, 8 | 887 | |||
| XM_017017506.1 | XP_016872995.1 | 7, 8 | 850 | |||
| XM_017017507.1 | XP_016872996.1 | 9, 10 | 1089 | |||
| XM_017017508.1 | XP_016872997.1 | 8, 9 | 964 | |||
| XM_017017509.1 | XP_016872998.1 | 6, 7 | 716 | |||
| GenBank mRNA | AK056362.1 | - | 6, 7 | 794 | ||
| AK092869.1 | - | 1, 2 | 1135 | |||
| AK301452.1 | - | 6, 7 | 788 | |||
| BC001073.2 | - | 6, 7 | 852 | 2820431 | ||
| BC001945.1 | - | 6, 7 | 852 | 2820431 | ||
| BC012856.1 | - | 6, 7 | 793 | 3871231 | ||
| BC017234.2 | - | 6, 7 | 801 | 3537381 | ||

HINFP
Genomic Map