siRNA Details
| Gene Symbol: IBTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300906.1 | NP_001287835.1 | 12, 13 | 2762 | ||
| NM_015525.3 | NP_056340.2 | 12, 13 | 2762 | |||
| XM_006715453.2 | XP_006715516.1 | 12, 13 | 3302 | |||
| XM_006715454.2 | XP_006715517.1 | 12, 13 | 3302 | |||
| GenBank mRNA | AB037838.2 | - | 11, 12 | 2486 | ||
| BC113696.1 | - | 11, 12 | 2291 | 8322748 | ||
| BC113698.1 | - | 11, 12 | 2291 | 8322750 | ||
| BC143745.1 | - | 11, 12 | 2292 | 9052259 | ||
| DQ005633.1 | - | 12, 13 | 2762 | |||
| DQ005634.1 | - | 12, 13 | 2762 | |||

IBTK
Genomic Map