siRNA Details
| Gene Symbol: ATP5J2-PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198879.1 | NP_001185808.1 | 5 | 945 | ||
| GenBank mRNA | AK295912.1 | - | 5 | 908 | ||
| Gene Symbol: PTCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015545.3 | NP_056360.2 | 4 | 912 | ||
| GenBank mRNA | AB014532.1 | - | 4 | 820 | ||
| AK001615.1 | - | 4 | 827 | |||
| AK291004.1 | - | 4 | 803 | |||
| BC003688.1 | - | 3 | 425 | |||
| BC080580.1 | - | 4 | 860 | 6645909 | ||
| BC103495.1 | - | 4 | 837 | 40011658 | ||
| BC103502.1 | - | 4 | 837 | 40011661 | ||
| BC103661.1 | - | 2 | 86 | |||

ATP5J2-PTCD1
Genomic Map