siRNA Details
| Gene Symbol: SUSD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015551.1 | NP_056366.1 | 5 | 1847 | ||
| XM_005265034.3 | XP_005265091.1 | 4 | 1395 | |||
| XM_017006137.1 | XP_016861626.1 | 5 | 1797 | |||
| GenBank mRNA | AB011099.1 | - | 5 | 1847 | ||
| AK295869.1 | - | 5 | 1442 | |||

SUSD5
Genomic Map