siRNA Details
| Gene Symbol: ZNF451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031623.2 | NP_001026794.1 | 13 | 3121 | ||
| NM_015555.2 | NP_056370.2 | 12 | 2977 | |||
| XM_011514460.1 | XP_011512762.1 | 12 | 2880 | |||
| XM_011514461.1 | XP_011512763.1 | 11 | 3496 | |||
| GenBank mRNA | AB011148.1 | - | 13 | 2940 | ||
| AK027805.1 | - | 4 | 410 | |||
| AK075174.1 | - | 4 | 824 | |||
| AK225806.1 | - | 13 | 2948 | |||
| AY055204.1 | - | 12 | 2978 | |||
| BC098382.1 | - | 13 | 2964 | 5295845 | ||
| BX648081.1 | - | 13 | 2967 | |||

ZNF451
Genomic Map