siRNA Details
| Gene Symbol: ZNF451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031623.2 | NP_001026794.1 | 10 | 2477 | ||
| NM_015555.2 | NP_056370.2 | 10 | 2477 | |||
| XM_005248994.1 | XP_005249051.1 | 10 | 2497 | |||
| XM_011514460.1 | XP_011512762.1 | 9 | 2236 | |||
| XM_011514461.1 | XP_011512763.1 | 8 | 2852 | |||
| XM_011514462.2 | XP_011512764.1 | 10 | 2497 | |||
| GenBank mRNA | AB011148.1 | - | 10 | 2296 | ||
| AK000983.1 | - | 1 | 983 | |||
| AK075174.1 | - | 1 | 180 | |||
| AK225806.1 | - | 10 | 2304 | |||
| AY055204.1 | - | 10 | 2478 | |||
| BC021712.2 | - | 10 | 2394 | 4821000 | ||
| BC098382.1 | - | 10 | 2320 | 5295845 | ||
| BX648081.1 | - | 10 | 2323 | |||

ZNF451
Genomic Map